Order Entry
Austria
Orders LinkContactUsLinkComponent
Vanillin

Vanillin

5 kg
Catalog # SIALW310727-5KG-K
Irritant
Supplier:   Merck
Vanillin
Catalog # SIALW310727-5KG-K
Supplier:  Merck
CAS Number:  121-33-5
Vanillin
Vanillin

5 kg

CAS Number:  121-33-5
Price
9 180,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:∼82 °C
  • Density:1.056 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ300
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Specifications

Specifications

  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.