Order Entry
Austria
Orders LinkContactUsLinkComponent
Vanillin

Vanillin

Glass bottle for solids, 100 g
Catalog # SIALW310727-100G-K
Irritant
Supplier:   Merck
Vanillin
Catalog # SIALW310727-100G-K
Supplier:  Merck
CAS Number:  121-33-5
Vanillin
Vanillin

Glass bottle for solids, 100 g

CAS Number:  121-33-5
Price
266,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:∼82 °C
  • Density:1.056 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ300
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications