Order Entry
Austria
Orders LinkContactUsLinkComponent
Hexanal, Sigma-Aldrich®

Hexanal, Sigma-Aldrich®

Glass bottle for solids, 100 g
Catalog # SIALW255726-100G-K
Flammable
Irritant
Supplier:   Merck
Hexanal, Sigma-Aldrich®
Catalog # SIALW255726-100G-K
Supplier:  Merck
CAS Number:  66-25-1
Hexanal, Sigma-Aldrich®

Glass bottle for solids, 100 g

CAS Number:  66-25-1
Price
223,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Hexylaldehyde, Caproaldehyde, Hexaldehyde, Aldehyde C6, Hexyl aldehyde
  • CAS Number:66-25-1
  • Formula:C₆H₁₂O
  • MDL Number:MFCD00007027
  • Molecular Weight:100.16 g/mol
  • Merck Index:13,01766
  • Boiling Pt:131 °C (1013 hPa)
  • Melting Pt:–56 °C
  • Density:0.81 g/cm³ (20 °C)
  • Flash Pt:25 °C
  • Storage Temperature:Refrigerator
  • UN:1207
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:200-624-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications