Hexanal, Sigma-Aldrich®
Glasflasche für Feststoffe, 1 kg
Catalog # SIALW255718-1KG-K
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Hexylaldehyde, Caproaldehyde, Hexaldehyde, Aldehyde C6, Hexyl aldehyde
- CAS Number:66-25-1
- Formula:C₆H₁₂O
- MDL Number:MFCD00007027
- Molecular Weight:100.16 g/mol
- Merck Index:13,01766
- Boiling Pt:131 °C (1013 hPa)
- Melting Pt:–56 °C
- Density:0,81 g/cm³ (20 °C)
- Flash Pt:25 °C
- Storage Temperature:Kühlschrank
- UN:1207
- ADR:3,III
- :M-Clarity™ Qualitätsstufe = MQ300
- EINECS:200-624-5
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



