Order Entry
Austria
Orders LinkContactUsLinkComponent
D-(-)-Mannitol, Sigma-Aldrich®

D-(-)-Mannitol, Sigma-Aldrich®

Glass bottle for solids, 5 kg
Catalog # SIALM4125-5KG
Supplier:   Merck
D-(-)-Mannitol, Sigma-Aldrich®
Catalog # SIALM4125-5KG
Supplier:  Merck
CAS Number:  69-65-8
D-(-)-Mannitol, Sigma-Aldrich®
D-(-)-Mannitol, Sigma-Aldrich®

Glass bottle for solids, 5 kg

CAS Number:  69-65-8
Price
664,00 €
Log in for availability
1 * 5 kg
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164…169 °C
  • Density:1.49 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:200-711-8

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.