Isosorbide dimethyl ether, Sigma-Aldrich®
Glass bottle for solids, 100 g
Catalog # SIAL247898-100G
Supplier: Merck
Isosorbide dimethyl ether, Sigma-Aldrich®
Catalog # SIAL247898-100G
Supplier: Merck
CAS Number: 5306-85-4
Jump to:
Product Details & Documents
Synonyms: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan
- CAS Number:5306-85-4
- Formula:C₈H₁₄O₄
- MDL Number:MFCD00064828
- Molecular Weight:174.20 g/mol
- :M-Clarity™ quality level = MQ100
- EINECS:226-159-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Cat. No.:SIAL247898-100G



