Order Entry
Austria
Orders LinkContactUsLinkComponent
 

pH buffers, colour coded, reference standards, Supelco®

4,00, ±0,01, Red, 25 °C, Plastic bottle, 6×500 ml, 6
  B5020-6X500ML
 :   Merck
pH buffers, colour coded, reference standards, Supelco®
  B5020-6X500ML
 :  Merck
 :  
pH buffers, colour coded, reference standards, Supelco®
pH buffers, colour coded, reference standards, Supelco®

4,00, ±0,01, Red, 25 °C, Plastic bottle, 6×500 ml, 6

 :  
 
237,00 €
   
1 * 6 ST
 

 : Standardised against N.I.S.T. standard reference materials

 

 

  • Pack type:
    Plastic bottle
  • Pk:
    6
  • Reference temperature:
    25 °C
  • Colour:
    Red
  • pH accuracy:
    ±0,01
  • Pk Info:
    6×500 ml
  • pH value:
    4,00
  • Cat. No.:
    B5020-6X500ML
  • M-Clarity quality level:
    MQ100