Order Entry
Austria
Orders LinkContactUsLinkComponent
Hexanal, Sigma-Aldrich®

Hexanal, Sigma-Aldrich®

Glass bottle, 2 mL
Catalog # SIAL115606-2ML
Flammable
Irritant
Supplier:   Merck
Hexanal, Sigma-Aldrich®
Catalog # SIAL115606-2ML
Supplier:  Merck
CAS Number:  66-25-1
Hexanal, Sigma-Aldrich®

Glass bottle, 2 mL

CAS Number:  66-25-1
Price
27,80 €
Log in for availability
1 * 2 mL
Product Details & Documents
Synonyms:  Hexylaldehyde, Caproaldehyde, Hexaldehyde, Aldehyde C6, Hexyl aldehyde
  • CAS Number:66-25-1
  • Formula:C₆H₁₂O
  • MDL Number:MFCD00007027
  • Molecular Weight:100.16 g/mol
  • Merck Index:13,01766
  • Boiling Pt:131 °C (1013 hPa)
  • Melting Pt:–56 °C
  • Density:0.81 g/cm³ (20 °C)
  • Flash Pt:25 °C
  • Storage Temperature:Refrigerator
  • UN:1207
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:200-624-5

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.