Orders LinkContactUsLinkComponent
Caps, ERLENMEYER-CAP
Caps, ERLENMEYER-CAP
Silver, 37-39 mm
Catalog # LUDI184115391
Supplier:   LUDI
Caps, ERLENMEYER-CAP
Catalog # LUDI184115391
Supplier:  LUDI
Caps, ERLENMEYER-CAP
Caps, ERLENMEYER-CAP
Silver, 37-39 mm
Price
54,50 €
Log in for availability
1 * 10 ST
Product Details & Documents
Aluminium, anodised.
  • Springs made of chromium-nickel steel
  • Springs clamp against the outer wall of the vessel
  • Autoclavable
Specifications
  • For:
    37-39 mm
  • Colour:
    Silver