Filter sets
Filter set 20, excitation BP 546/12, beam splitter FT 560, emission BP 575-640
Catalog # ZEIS4880209901000
Supplier: ZEISS
Jump to:
Product Details & Documents
Specifications
- Description:Filter set 20, excitation BP 546/12, beam splitter FT 560, emission BP 575-640
- Seq:05