ContactUsLinkComponent
HOMOGENIZER PRECELLYS 24 DUAL homogenizers dedicated to the grinding, lysis and homogenization of biological samples prior to DNA, RNA and proteins xtraction. 12 x 2mL/0.5mL OR 6 x 7mL tubes holder, 500rpm to 6500rpm. for soft and hard tissues, plant 1 * 1 items
Catalog # 432-3757
Supplier:  Bertin Instruments
HOMOGENIZER PRECELLYS 24 DUAL homogenizers dedicated to the grinding, lysis and homogenization of biological samples prior to DNA, RNA and proteins xtraction. 12 x 2mL/0.5mL OR 6 x 7mL tubes holder, 500rpm to 6500rpm. for soft and hard tissues, plant 1 * 1 items
Catalog # 432-3757
Supplier:  Bertin Instruments
Frequently Bought Together