Orders LinkContactUsLinkComponent
Camera adapters
Camera adapters
0,65×C-mount camera adapter for ²/₃" chip sensors
Catalog # 630-1945
Supplier:   MOTIC
Camera adapters
Catalog # 630-1945
Supplier:  MOTIC
Camera adapters
Camera adapters
0,65×C-mount camera adapter for ²/₃" chip sensors
Price
159,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    0,65×C-mount camera adapter for ²/₃" chip sensors
  • Seq:
    04