Orders LinkContactUsLinkComponent
Inverted microscopes, AE2000
Inverted microscopes, AE2000
Widefield eyepiece N-WF 15×/16 mm with diopter adjustment
  630-1456
 :   MOTIC
Inverted microscopes, AE2000
  630-1456
 :  MOTIC
Inverted microscopes, AE2000
Inverted microscopes, AE2000
Widefield eyepiece N-WF 15×/16 mm with diopter adjustment
118,00 €
1 * 1 ST
AE2000 inverted microscopes are designed for cell culture applications.
  • Automatic turn off after 15 m of non use
  • RoHs compliant lead-free objectives
  • Robust manufacturing with small footprint
Suitable for microbiology and clinical labs, especially for routine cell inspections.
Ordering information: Special sets including microscope, application-specific accessories and three objectives (4×, Ph 10×, LWD Ph 20×) are available as well.
Delivery information: Additionnal phase contrast objectives are also available. LED modules can be ordered separately.
  • Magnification:
    15×/16 mm
  • Description:
    Widefield eyepiece N-WF 15×/16 mm with diopter adjustment
  • Seq:
    201
  • Cat. No.:
    630-1456
  • Condenser:
    0,30 N.A.
  • Nosepiece:
    Quadruple
Accessories
  •  
    Description
  • 630-1385
    Phase slider, 3 positions
    1 * 1 ST - 146,00 €
  •  
    Description
  • 630-1373
    Stage holder for 35 mm Petri dishes
    1 * 1 ST - 63,00 €
  •  
    Description
  • 630-1383
    Ground glass filter (Ø = 45 mm)
    1 * 1 ST - 15,00 €
  •  
    Description
  • 630-1386
    Neutral density filter (Ø = 45 mm)
    1 * 1 ST - 44,00 €
  •  
    Description
  • 630-1502
    1× C-mount camera adapter (no lens)
    1 * 1 ST - 138,00 €
  •  
    Description
  • 630-1381
    Phase ring Ph1
    1 * 1 ST - 38,00 €
  •  
    Description
  • 630-1379
    Condenser 0,30 N.A. WD: 72 mm
    1 * 1 ST - 308,00 €
  •  
    Description
  • 630-1380
    Phase ring Ph0
    1 * 1 ST - 38,00 €
  •  
    Description
  • 630-1376
    Auxiliary stages (one pair)
    1 * 1 ST - 64,00 €
  •  
    Description
  • 630-1387
    Metal stage insert
    1 * 1 ST - 40,00 €
  •  
    Description
  • 630-1503
    0,5×C-mount camera adapter for ¹/₃ and ½" chip sensors
    1 * 1 ST - 159,00 €
  •  
    Description
  • 630-1374
    Stage holder for 54 mm Petri dishes
    1 * 1 ST - 63,00 €
  •  
    Description
  • 630-1320
    LED module 4500 K ±300 K
    1 * 1 ST - 90,00 €
  •  
    Description
  • 630-1319
    LED module 6000 K ±300 K
    1 * 1 ST - 90,00 €
  •  
    Description
  • 630-1378
    Condenser 0,40 N.A (WD = 53 mm)
    1 * 1 ST - 550,00 €
  •  
    Description
  • 630-1500
    Attachable stage with well plate holder, 128×86 mm
    1 * 1 ST - 707,00 €
  •  
    Description
  • 630-1377
    Glass stage insert
    1 * 1 ST - 19,00 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.