Order Entry
Austria
Orders LinkContactUsLinkComponent
Sample Bags without Closure

Sample Bags without Closure

600×400 mm, 0,20 mm
Catalog # SCAC2092
Sample Bags without Closure
Catalog # SCAC2092
Sample Bags without Closure
Sample Bags without Closure

600×400 mm, 0,20 mm

Price
1 760,00 €
Log in for availability
1 * 1.400 ST
Product Details & Documents

Bags made from PE transparent, for the storage or transport of samples.

Available in several sizes.

 : Supplied without closure ties.

Specifications
  • L×W:
    600×400 mm
  • Seq:
    13
  • Thickness:
    0,20 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.