Orders LinkContactUsLinkComponent
Objectives
Objectives
A-Planachromatic, M27, N.A. of 0,25, WD of 4,5 mm
Catalog # ZEIS4210409900000
Supplier:   ZEISS
Objectives
Catalog # ZEIS4210409900000
Supplier:  ZEISS
Objectives
Objectives
A-Planachromatic, M27, N.A. of 0,25, WD of 4,5 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    10×
  • Description:
    A-Planachromatic, M27, N.A. of 0,25, WD of 4,5 mm
  • Seq:
    05