Forceps, precision
Serrated handle, curved, round, broad, flat tips, 35B, 120 mm
Catalog # OUTI35B
Supplier: OUTILS RUBIS
Springe zu:
Product Details & Documents
Stainless steel. Forceps with very precisely machined tips. Ideal for applications that require a high degree of precision, including microscopy, anatomy, biology, microbiology, gene technology and electron microscopy.
- Acid-resistant
- Resistant against hydrogen fluoride and nitric acid
- 100% anti-magnetic
Dokumente
Spezifikationen
- Type:35B
- Description:Serrated handle, curved, round, broad, flat tips
- Length:120 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



