Order Entry
Austria
Orders LinkContactUsLinkComponent
Forceps, precision

Forceps, precision

Straight, extra wide flat tips, 35A, 120 mm
Catalog # 232-0209
Supplier:   OUTILS RUBIS
Forceps, precision
Catalog # 232-0209
Supplier:  OUTILS RUBIS
Forceps, precision
Forceps, precision

Straight, extra wide flat tips, 35A, 120 mm

Price
31,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Forceps with very precisely machined tips. Ideal for applications that require a high degree of precision, including microscopy, anatomy, biology, microbiology, gene technology and electron microscopy.

  • Acid-resistant
  • Resistant against hydrogen fluoride and nitric acid
  • 100% anti-magnetic

Documents

Specifications
  • Type:
    35A
  • Description:
    Straight, extra wide flat tips
  • Length:
    120 mm