Ammonia solution 25%, TECHNICAL
Plastic drum, 17 kg
Anbieter: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Boiling Pt:32…37.7 °C (1013 hPa)
- Melting Pt:–60…–57.5 °C
- Density:0.907 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:2672
- ADR:8,III
Documents
Certificates
Search by Lot Number5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Packaging Information
P20_VWRC21191.364_SPEC_EN.pdf P20_VWRC21191.450_SPEC_EN.pdf spec_VWRC21191.364 spec_VWRC21191.364 spec_VWRC21191.450
Product specification
Mehr
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Assay:Min. 24 - 26 %
Specifications
- Pack type:Plastic drum
- Pk:17 kg



