ContactUsLinkComponent
A specific mRNA is reverse transcribed to double-stranded DNA. This DNA product is then amplified by PCR. This reaction demonstrates the mode of replication of HIV, which contains reverse transcriptase. This experiment is the first introduction of a 1 * 1 KIT
Catalog # EDVO335
Supplier:  EDVOTEK
A specific mRNA is reverse transcribed to double-stranded DNA. This DNA product is then amplified by PCR. This reaction demonstrates the mode of replication of HIV, which contains reverse transcriptase. This experiment is the first introduction of a 1 * 1 KIT
Catalog # EDVO335
Supplier:  EDVOTEK
Frequently Bought Together