ContactUsLinkComponent
Teach your students about synthetic biology with this exciting and exclusive lab! Students use PCR to amplify the coding sequence of the BSMT1 enzyme. This interesting enzyme is responsible for the formation of methyl salicylate, a chemical with a s 1 * 1 KIT
Catalog # EDVO331
Supplier:  EDVOTEK
Teach your students about synthetic biology with this exciting and exclusive lab! Students use PCR to amplify the coding sequence of the BSMT1 enzyme. This interesting enzyme is responsible for the formation of methyl salicylate, a chemical with a s 1 * 1 KIT
Catalog # EDVO331
Supplier:  EDVOTEK
Frequently Bought Together