Trione Ninhydrin Reagenz 1 * 1 items
Catalog # MSPPT200
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
N,N-Dimethylhydrazine
Supplier: Merck
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Diphosphorus pentoxide, Supelco®
Supplier: Merck
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Methyl red solution in ethanol acid-base indicator, Fluka™
Supplier: Honeywell Chemicals
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1 L
Supplier: PanReac AppliChem
1,2,4-Trichlorobenzene ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals