Filter sets
Filter set 09, excitation BP 450 - 490, beam splitter FT 510, emission LP 515
Artikelnr. ZEIS4880099901000
Anbieter: ZEISS
Jump to:
Product Details & Documents
Specifications
- Description:Filter set 09, excitation BP 450 - 490, beam splitter FT 510, emission LP 515
- Seq:02