Orders LinkContactUsLinkComponent
Drum funnels
Drum funnels
Catalog # NEWPDRM452-BK
Supplier:   NEW PIG
Drum funnels
Catalog # NEWPDRM452-BK
Supplier:  NEW PIG
Drum funnels
Drum funnels
Price
67,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Thick, double-wall polyethylene construction resists UV rays, corrosion and most chemicals.
  • Allows access to ³/₄" bung for use of liquid-level gauge
  • Level inside surface allows easy draining of containers, parts or filters
  • Zigzag ridges prevent splashes and catch parts or tools
These large diameter funnels covers entire drum top to minimize drips and spills.
Accessories
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Pop-up drum fill gauge, prevent overfilling
    Pop-up drum fill gauge, prevent overfilling
    NEWPDRM245
    127 mm
    Log in for availability
    1 * 1 ST - 26,50 €
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Pop-up drum fill gauge, prevent overfilling
    Pop-up drum fill gauge, prevent overfilling
    NEWPDRM317
    279,4 mm
    Log in for availability
    1 * 1 ST - 30,30 €