ContactUsLinkComponent
ChemiDoc-It 600 System. With OptiChemi HR camera: 16 bit monochrome, 16 File Bit Depth (A/D), digital camera and mount. 2184 x 1471 Kodak chip with binning capability (3.2 Mp). Peltier cooling to -60C from ambient (please inquire for deeper cooling 1 * 1 items
Catalog # 733-2026
ChemiDoc-It 600 System. With OptiChemi HR camera: 16 bit monochrome, 16 File Bit Depth (A/D), digital camera and mount. 2184 x 1471 Kodak chip with binning capability (3.2 Mp). Peltier cooling to -60C from ambient (please inquire for deeper cooling 1 * 1 items
Catalog # 733-2026
Frequently Bought Together