BioSpectrum 800 2D Imaging System. With MegaCam camera: 16 bit monochrome, digital camera and mount. 3326 x 2504 resolution with bin ning capability (8.3 MP). Peltier cooling to -50C from ambient. Camera mount and fixed lens 28mm, f/1.8, close up dio 1 * 1 items
Catalog # UVPA97-0475-02
Supplier: UVP ULTRA VIOLET PRODUCTS
Frequently Bought Together