ContactUsLinkComponent
BioSpectrum 600 System. With OptiChemi HR camera: 16 bit monochrome, digital camera and mount. 2184 x 1472 Kodak chip with binning c apability (3.2 Mp). Peltier cooling to -60C from ambient (please inquire for deeper cooling options). Camera mount an 1 * 1 items
Catalog # UVPA97-0410-02
BioSpectrum 600 System. With OptiChemi HR camera: 16 bit monochrome, digital camera and mount. 2184 x 1472 Kodak chip with binning c apability (3.2 Mp). Peltier cooling to -60C from ambient (please inquire for deeper cooling options). Camera mount an 1 * 1 items
Catalog # UVPA97-0410-02
Frequently Bought Together