[DE]Eichlösung 300 mosm 5 x 10 AMP 1 * 50 Ampoul
Catalog # MSPPVO-4-815.01
Supplier: MARKET SOURCE PART PROCESS
[DE]Eichlösung 300 mosm 5 x 10 AMP 1 * 50 Ampoul
Catalog # MSPPVO-4-815.01
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals