Chiller UC015-H air-cooled B -10-100C Cooling capacity (0C) 1kW Pump connection 3/4 1 * 1 items
461-0995
: HUBER
Chiller UC015-H air-cooled B -10-100C Cooling capacity (0C) 1kW Pump connection 3/4 1 * 1 items
461-0995
: HUBER
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection