Orders LinkContactUsLinkComponent
Filter sets
Filter set 38 (high efficiency), excitation BP 470/40 (HE), beam splitter FT 495 (HE), emission BP 525/50 (HE)
  ZEIS4890389901000
 :   ZEISS
Filter sets
  ZEIS4890389901000
 :  ZEISS
Filter sets
Filter set 38 (high efficiency), excitation BP 470/40 (HE), beam splitter FT 495 (HE), emission BP 525/50 (HE)
1 * 1 ST
  • Description:
    Filter set 38 (high efficiency), excitation BP 470/40 (HE), beam splitter FT 495 (HE), emission BP 525/50 (HE)
  • Seq:
    09