Universal socks
Universal sorbent snakes, 15 per pack, 75×1200 mm
Catalog # MSGA9954
Supplier: asecos
Jump to:
Product Details & Documents
Universal socks for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
- Optimal for long term use
- Capacity up to 16 times of its own weight
- High rib and rub resistance
For containment and absorption of larger leaks.
Specifications
- Description:Universal sorbent snakes
- Packed:15 per pack
- Ø×L:75×1200 mm