TPN 380, K-Type surface probe, up to 800°C, Ø 15mm, L 300mm, 1m silicone cable fixed 1 * 1 items
EBRO1341-0720
: EBRO
TPN 380, K-Type surface probe, up to 800°C, Ø 15mm, L 300mm, 1m silicone cable fixed 1 * 1 items
EBRO1341-0720
: EBRO
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK