TPN 920, K-Type magnetic surface probe, up to 400°C, 1m fixed thermo-wire 1 * 1 items
Catalog # EBRO1341-0645
Supplier: EBRO
TPN 920, K-Type magnetic surface probe, up to 400°C, 1m fixed thermo-wire 1 * 1 items
Catalog # EBRO1341-0645
Supplier: EBRO
Frequently Bought Together
VWR®, Probes for Precise PT1000 Thermometer TD23
Supplier: VWR Collection
VWR®, Single-Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
Single-channel pipettes, mechanical, fixed / variable volume, Finnpipette™ F2
Supplier: Thermo Fisher Scientific
VWR® VAPOUR-Line <i>Lite </i>II Autoclaves
Supplier: VWR Collection
COVER - SGLE WFR 150MM H22-601-0615 1 * 100 ST
Supplier: ENTEGRIS
Accessories for 5702 / 5702 R / 5702 RH
Supplier: EPPENDORF
Bath fluids
Supplier: HUBER
Rotors for Hettich Centrifuges
Supplier: Hettich
Accessories for pipette controller, BRAND accu-jet® S
Supplier: Brand
Cleanroom Waste Receptacles
Supplier: Palbam
VWR® Traceable®, Thermometers for Refrigerators and Freezers
Supplier: VWR Collection
Accessories for thermometers smart and wireless probes
Supplier: Testo