Order Entry
Austria
Orders LinkContactUsLinkComponent
Baumé hydrometers

Baumé hydrometers

65 - 75, 0,1, 300 mm
  ALLA0050FG075/15QP
Baumé hydrometers
  ALLA0050FG075/15QP
Baumé hydrometers
Baumé hydrometers

65 - 75, 0,1, 300 mm

 
50,80 €
   
1 * 1 ST
 

Baumé hydrometers calibrated at 15 °C.

 
  • Length:
    300 mm
  • Seq:
    30
  • Division:
    0,1
  • Range:
    65 - 75