Orders LinkContactUsLinkComponent
FlashGel® camera
FlashGel® camera, includes camera, hood enclosure, USB cable and software
Catalog # 730-1346
Supplier:   LONZA
FlashGel® camera
Catalog # 730-1346
Supplier:  LONZA
FlashGel® camera
FlashGel® camera
FlashGel® camera, includes camera, hood enclosure, USB cable and software
Price
1 350,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The FlashGel® camera captures data from the FlashGel® system, allowing complete separation, recovery and documentation safely, at the bench, in minutes. This simple digital camera in an enclosed hood connects directly to a laptop, PC or PDA via USB.
  • Complete gel run and image capture in five minutes
  • Photograph gels at the bench, without UV light
  • Sharp, clear, high-resolution images
  • Simple user interface
  • Optimised exposure for FlashGel® cassettes
Hood (W×L×H): 100×110×160 mm
Image file type: .JPG, .TIFF, .PDF
Specifications
  • Description:
    FlashGel® camera, includes camera, hood enclosure, USB cable and software
  • Seq:
    01