Order Entry
Austria
Orders LinkContactUsLinkComponent
Pasteur pipettes, Corning®

Pasteur pipettes, Corning®

Non plugged, borosilicate glass, 6,5 mm, 229 mm, -, 200/pack
Catalog # 612-5200
Supplier:   Corning
Pasteur pipettes, Corning®
Catalog # 612-5200
Supplier:  Corning
Pasteur pipettes, Corning®
Pasteur pipettes, Corning®

Non plugged, borosilicate glass, 6,5 mm, 229 mm, -, 200/pack

Price
85,40 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Soda-lime or borosilicate glass.

  • Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
  • Features a constriction one inch below the top to facilitate plugging
  • Available in convenient bulk packs

These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, haematology and serology studies, as well as blood bank and clinical chemistry procedures.

Specifications
  • Ext.Ø:
    6,5 mm
  • Packed:
    200/pack
  • Sterile:
    -
  • Description:
    Non plugged, borosilicate glass
  • Length:
    229 mm
  • Seq:
    04