Orders LinkContactUsLinkComponent
Microscope slides, Corning®
Microscope slides, Corning®
Frosted on one side slides, Colourless, 02, Glass, 75×25 mm
Catalog # 734-4039
Supplier:   Corning
Microscope slides, Corning®
Catalog # 734-4039
Supplier:  Corning
Microscope slides, Corning®
Microscope slides, Corning®
Frosted on one side slides, Colourless, 02, Glass, 75×25 mm
Price
509,00 €
Log in for availability
1 * 1.440 ST
Product Details & Documents
These slides are either frosted on one side, on both sides, or plain.
  • Clarity reinforced water-white glass
  • Frosted slides are sandblasted, easy to write on and highly legible
  • Each slide is inspected for chips and rough edge before being shipped
Documents
Specifications
  • No. of wells:
    02
  • Colour:
    Colourless
  • Description:
    Frosted on one side slides
  • L×W:
    75×25 mm
  • Material:
    Glass
  • Seq:
    02