Orders LinkContactUsLinkComponent
Control Cables
Control Cables
Control cable to connect Com.G@te® AIF to cable end open, 3 m
Catalog # HUBE9353
Supplier:   HUBER
Control Cables
Catalog # HUBE9353
Supplier:  HUBER
Control Cables
Control Cables
Control cable to connect Com.G@te® AIF to cable end open, 3 m
Price
89,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
A range of control cables is available for USB, RS232, RS485 or analogue interfaces (AIF). Cables are also available for external control signal (ECS), a potentialfree contact (POKO) or for an external float switch (LEVEL).
Specifications
  • Description:
    Control cable to connect Com.G@te® AIF to cable end open
  • Length:
    3 m
  • Seq:
    05