Order Entry
Austria
Orders LinkContactUsLinkComponent
Bath fluids

Bath fluids

SilOil, 20...275 °C, 300 °C, 50 mm²/s, 5 L
Catalog # HUBE6157
Supplier:   HUBER
Bath fluids
Catalog # HUBE6157
Supplier:  HUBER
CAS Number:  
Bath fluids
Bath fluids

SilOil, 20...275 °C, 300 °C, 50 mm²/s, 5 L

CAS Number:  
Price
450,00 €
Log in for availability
1 * 5 L
Product Details & Documents

Thermal fluids with a wide range of operating temperatures and good thrmodynamic properties for use in circulators and heating systems where water is not appropriate.

All viscosities values are dynamic visosities measured at 25 °C except DW-Therm® which was measured at 20 °C.

Caution: * –20...+235 °C for closed system applications

Use biocides safely. Always read the label and product information before use

Specifications

Specifications

  • Pk:
    5 L
  • Flash point:
    300 °C
  • Temperature range:
    20...275 °C
  • Type:
    SilOil
  • Viscosity:
    50 mm²/s

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.