[DE]Newtonsche Normalprobe 5000 AW 1 * 100 ml
MSPP5000AW-100ML
Frequently Bought Together
o-Xylene anhydrous, 97 % 1 * 2 L
: Merck
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
tert-Butanol ACS
: Thermo Fisher Scientific
N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal®
: Thermo Fisher Scientific
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Ethanolamine, Sigma-Aldrich®
: Merck
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Methyl ethyl ketone ACS
: Thermo Fisher Scientific
Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™
: Honeywell Chemicals