Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 02, excitation G 365, beam splitter FT 395, emission LP 420
Catalog # ZEIS4880029901000
Supplier:   ZEISS
Filter sets
Catalog # ZEIS4880029901000
Supplier:  ZEISS
Filter sets
Filter set 02, excitation G 365, beam splitter FT 395, emission LP 420
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Filter set 02, excitation G 365, beam splitter FT 395, emission LP 420
  • Seq:
    01