Drum funnels
Catalog # NEWPDRM672-BK
Supplier: NEW PIG
Jump to:
Product Details & Documents
Thick, double-wall polyethylene construction resists UV rays, corrosion and most chemicals.
- Allows access to ³/₄" bung for use of liquid-level gauge
- Level inside surface allows easy draining of containers, parts or filters
- Zigzag ridges prevent splashes and catch parts or tools
These large diameter funnels covers entire drum top to minimize drips and spills.
Accessories
Product Information
- Catalog #LengthAvailabilityPrice
- Pop-up drum fill gauge, prevent overfillingPop-up drum fill gauge, prevent overfillingNEWPDRM245127 mmLog in for availability1 * 1 ST - 26,50 €
Product Information
- Catalog #LengthAvailabilityPrice
- Pop-up drum fill gauge, prevent overfillingPop-up drum fill gauge, prevent overfillingNEWPDRM317279,4 mmLog in for availability1 * 1 ST - 30,30 €