Order Entry
Austria
Orders LinkContactUsLinkComponent
Buckets

Buckets

10 L, 295/170 mm, 270 mm
Catalog # COLDBUC4010G
Buckets
Catalog # COLDBUC4010G
Buckets
Buckets

10 L, 295/170 mm, 270 mm

Price
184,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, 304 grade.

  • Very good chemical resistance, very good thermal resistance
  • High quality, very stable
  • An optional lid is available which is extended to cover the spout

These buckets are polished throughout, graduated in 1 litre intervals and come complete with swing handle. Skirtbands are standard. They have crevice and pit free interiors and are polished throughout.

Specifications
  • Capacity:
    10 L
  • Height:
    270 mm
  • Ø top/Ø bottom:
    295/170 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.