Microscope slides
Microscope slides, 1,0 - 1,2 mm, 76×26 mm
SMITMC2951B/76X26
These slides are made of glass with ground edges.
- Description:Microscope slides
- L×W:76×26 mm
- Seq:001
- Thickness:1,0 - 1,2 mm
These slides are made of glass with ground edges.