Order Entry
Austria
Orders LinkContactUsLinkComponent
Reference electrode filling solutions

Reference electrode filling solutions

Potassium chloride (KCl) 3 mol/l, AgCl saturated, for Ag/AgCl reference systems, 25 mL
Catalog # METT51343184
Supplier:   Mettler - Toledo
Reference electrode filling solutions
Catalog # METT51343184
Supplier:  Mettler - Toledo
CAS Number:  
Reference electrode filling solutions
Reference electrode filling solutions

Potassium chloride (KCl) 3 mol/l, AgCl saturated, for Ag/AgCl reference systems, 25 mL

CAS Number:  
Price
12,90 €
Log in for availability
1 * 25 mL
Product Details & Documents

FRISCOLYT-B® (84,4% glycerol, 11% KCl and 4,6% H₂O) is for low temperature samples.

Specifications

Specifications

  • Pk:
    25 mL
  • Description:
    Potassium chloride (KCl) 3 mol/l, AgCl saturated, for Ag/AgCl reference systems

Specifications