Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Reference electrode filling solutions

Potassium nitrate (KNO₃) 1 mol/l, bridge electrolyte, for DX series ISE half-cells, 25 mL
Catalog # METT51343182
Supplier:   Mettler - Toledo
Reference electrode filling solutions
Catalog # METT51343182
Supplier:  Mettler - Toledo
CAS Number:  
Reference electrode filling solutions
Reference electrode filling solutions

Potassium nitrate (KNO₃) 1 mol/l, bridge electrolyte, for DX series ISE half-cells, 25 mL

CAS Number:  
Price
12,90 €
Log in for availability
1 * 25 mL
Product Details & Documents

FRISCOLYT-B® (84,4% glycerol, 11% KCl and 4,6% H₂O) is for low temperature samples.

Specifications

Specifications

  • Pk:
    25 mL
  • Description:
    Potassium nitrate (KNO₃) 1 mol/l, bridge electrolyte, for DX series ISE half-cells

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.