Order Entry
Austria
Orders LinkContactUsLinkComponent
Quality control test suspensions

Quality control test suspensions

Escherichia coli, 2,5 mL
Catalog # NAMSGP25-09
Supplier:   CROSSTEX
Quality control test suspensions
Catalog # NAMSGP25-09
Supplier:  CROSSTEX
CAS Number:  
Quality control test suspensions

Escherichia coli, 2,5 mL

CAS Number:  
Price
193,00 €
Log in for availability
1 * 2,5 mL
Product Details & Documents

The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.

  • 2,5 ml pro vial
  • 25 dose
Specifications

Specifications

  • Pk:
    2,5 mL
  • Description:
    Escherichia coli

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.