Orders LinkContactUsLinkComponent
Quality control test suspensions
Quality control test suspensions
Pseudomonas aeruginosa, 2,5 mL
Catalog # NAMSGP25-07
Quality control test suspensions
Catalog # NAMSGP25-07
Supplier:  
CAS Number:  
Quality control test suspensions
Pseudomonas aeruginosa, 2,5 mL
CAS Number:  
Discontinued
Product Details & Documents
The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.
  • 2,5 ml pro vial
  • 25 dose
Spezifikationen
Spezifikationen
  • Pk:
    2,5 mL
  • Description:
    Pseudomonas aeruginosa
Spezifikationen