Order Entry
Austria
Orders LinkContactUsLinkComponent
Quality control test suspensions

Quality control test suspensions

Candida albicans, 2,5 mL
Catalog # NAMSGP25-03
Supplier:   CROSSTEX
Quality control test suspensions
Catalog # NAMSGP25-03
Supplier:  CROSSTEX
CAS Number:  
Quality control test suspensions

Candida albicans, 2,5 mL

CAS Number:  
Price
193,00 €
Log in for availability
1 * 2,5 mL
Product Details & Documents

The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.

  • 2,5 ml pro vial
  • 25 dose
Specifications

Specifications

  • Pk:
    2,5 mL
  • Description:
    Candida albicans

Specifications