IsoFlow Sheath Fluid for Cytometer FC500 1 * 10 L
Catalog # BECL8546859
Supplier: Beckman Coulter
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
CytoFLEX Ready to Use Daily QC Fluorospheres 1 * 20 mL
Supplier: Beckman Coulter