ContactUsLinkComponent
Thermometer, general purpose, simple type, mercury filled, measuring range:-10:50/0.5C, length:300mm, diameter:6-7mm, 76mm immersio n, solid stem, yellow backed, with ring at top, durable pigment. 1 * 1 items
Catalog # 620-0074
Supplier:  Amarell
Thermometer, general purpose, simple type, mercury filled, measuring range:-10:50/0.5C, length:300mm, diameter:6-7mm, 76mm immersio n, solid stem, yellow backed, with ring at top, durable pigment. 1 * 1 items
Catalog # 620-0074
Supplier:  Amarell
Frequently Bought Together